Assay Details
| Gene Symbol: FOXN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261043.1 | NP_001247972.1 | 777 | 130 | ||
| XM_015112307.2 | XP_014967793.1 | 966 | 130 | |||
| XM_015112309.2 | XP_014967795.1 | 1100 | 130 | |||
| XM_015112310.2 | XP_014967796.1 | 866 | 130 | |||
| XM_015112311.2 | XP_014967797.1 | 864 | 130 | |||
| XM_028831621.1 | XP_028687454.1 | 1143 | 130 | |||
| XM_028831622.1 | XP_028687455.1 | 1325 | 130 | |||
| XM_028831623.1 | XP_028687456.1 | 1338 | 130 | |||
| XM_028831624.1 | XP_028687457.1 | 849 | 130 | |||
| GenBank mRNA | JU334307.1 | - | 866 | 130 | ||
| JU334308.1 | - | 866 | 130 | |||
| JU475413.1 | - | 777 | 130 | |||
| JU475414.1 | - | 777 | 130 | |||
More Information
Set Membership: |
|

FOXN2
Genomic Map