Assay Details
| Gene Symbol: KCNJ3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261696.1 | NP_001248625.1 | 865 | 96 | ||
| XM_028830410.1 | XP_028686243.1 | 966 | 96 | |||
| GenBank mRNA | JU320252.1 | - | 865 | 96 | ||
| JV601784.1 | - | 912 | 96 | |||
More Information
Set Membership: |
|

KCNJ3
Genomic Map