Assay Details
| Gene Symbol: LAMP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260808.1 | NP_001247737.1 | 728 | 72 | ||
| XM_015128101.2 | XP_014983587.1 | 809 | 72 | |||
| XM_015128102.2 | XP_014983588.1 | 809 | 72 | |||
| GenBank mRNA | JU475560.1 | - | 696 | 72 | ||
| JU475561.1 | - | 696 | 72 | |||
| JV043335.1 | - | 728 | 72 | |||
| JV047036.1 | - | 728 | 72 | |||
More Information
Set Membership: |
|

LAMP2
Genomic Map