Assay Details
| Gene Symbol: LRPAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001085674.4 | XP_001085674.2 | 1406 | 151 | ||
| GenBank mRNA | JU334497.1 | - | 1035 | 151 | ||
| JU475589.1 | - | 1025 | 151 | |||
| JV544937.1 | - | 1029 | 151 | |||
| JV600281.1 | - | 1031 | 151 | |||
| JV684044.1 | - | 1037 | 151 | |||
More Information
Set Membership: |
|

LRPAP1
Genomic Map