Assay Details
| Gene Symbol: MNDA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001117129.4 | XP_001117129.2 | 1392 | 125 | ||
| XM_015112693.2 | XP_014968179.2 | 1425 | 125 | |||
| GenBank mRNA | JU334573.1 | - | 1365 | 125 | ||
| JU475658.1 | - | 1363 | 125 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

MNDA
Genomic Map