Assay Details
| Gene Symbol: NUBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015125608.2 | XP_014981094.2 | 620 | 126 | ||
| XM_028841403.1 | XP_028697236.1 | 620 | 126 | |||
| GenBank mRNA | JU334627.1 | - | 378 | 126 | ||
| JU475693.1 | - | 390 | 126 | |||
| JU475694.1 | - | 390 | 126 | |||
| JV047143.1 | - | 369 | 126 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

NUBP1
Genomic Map