Assay Details
| Gene Symbol: PCNA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266090.1 | NP_001253019.1 | 897 | 124 | ||
| XM_015149367.2 | XP_015004853.1 | 840 | 124 | |||
| XM_015149368.2 | XP_015004854.1 | 976 | 124 | |||
| GenBank mRNA | JU334791.1 | - | 904 | 124 | ||
| JU475825.1 | - | 935 | 124 | |||
| JU475826.1 | - | 917 | 124 | |||
| JV047251.1 | - | 897 | 124 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

PCNA
Genomic Map