Assay Details
| Gene Symbol: RDH5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001110917.4 | XP_001110917.1 | 1452 | 73 | ||
| XM_015151915.2 | XP_015007401.1 | 1447 | 73 | |||
| GenBank mRNA | JU478298.1 | - | 780 | 73 | ||
More Information
Set Membership: |
|

RDH5
Genomic Map