Assay Details
| Gene Symbol: LOC100425930 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028841316.1 | XP_028697149.1 | 91 | 142 | ||
| Gene Symbol: LOC699599 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028841314.1 | XP_028697147.1 | 76 | 142 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_028841314.1, XM_028841316.1 transcripts.
Set Membership: |
|

LOC100425930
Genomic Map