Assay Details
| Gene Symbol: LOC114675624 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028842839.1 | XP_028698672.1 | 679 | 123 | ||
| Gene Symbol: CASK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015127187.2 | XP_014982673.1 | 2007 | 123 | ||
| XM_015127188.2 | XP_014982674.1 | 2008 | 123 | |||
| XM_015127189.2 | XP_014982675.1 | 1989 | 123 | |||
| XM_015127190.2 | XP_014982676.1 | 1989 | 123 | |||
| XM_015127191.2 | XP_014982677.1 | 1855 | 123 | |||
| XM_015127192.2 | XP_014982678.1 | 1970 | 123 | |||
More Information
Set Membership: |
|

LOC114675624
Genomic Map