Assay Details
| Gene Symbol: TNFSF13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205136.3 | NP_001192065.2 | 1250 | 94 | ||
| XM_015118488.2 | XP_014973974.1 | 789 | 94 | |||
| XM_015118489.2 | XP_014973975.1 | 1250 | 94 | |||
| XM_015118490.2 | XP_014973976.1 | 1202 | 94 | |||
| XM_015118491.2 | XP_014973977.1 | 556 | 94 | |||
| GenBank mRNA | JU336905.1 | - | 553 | 94 | ||
| JV597155.1 | - | 810 | 94 | |||
| JV668527.1 | - | 837 | 94 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

TNFSF13
Genomic Map