Assay Details
| Gene Symbol: TNFRSF14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001043357.1 | NP_001036822.1 | 298 | 129 | ||
| GenBank mRNA | DQ851094.1 | - | 298 | 129 | ||
| JU495558.1 | - | 580 | 129 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

TNFRSF14
Genomic Map