Assay Details
| Gene Symbol: RNF8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001265829.1 | NP_001252758.1 | 364 | 133 | ||
| XM_015136129.2 | XP_014991615.1 | 446 | 133 | |||
| XM_015136130.2 | XP_014991616.1 | 286 | 133 | |||
| XM_015136131.2 | XP_014991617.1 | 446 | 133 | |||
| XM_028846598.1 | XP_028702431.1 | 446 | 133 | |||
| GenBank mRNA | JU336991.1 | - | 392 | 133 | ||
| JU477714.1 | - | 364 | 133 | |||
| JV048627.1 | - | 391 | 133 | |||
| JV048628.1 | - | 391 | 133 | |||
| JV597682.1 | - | 414 | 133 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

RNF8
Genomic Map