Assay Details
| Gene Symbol: AIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194313.1 | NP_001181242.1 | 234 | 101 | ||
| XM_015113521.2 | XP_014969007.1 | 286 | 101 | |||
| GenBank mRNA | JU337002.1 | - | 226 | 101 | ||
| JV045056.1 | - | 219 | 101 | |||
| JV597797.1 | - | 231 | 101 | |||
| JV726470.1 | - | 224 | 101 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

AIP
Genomic Map