Assay Details
| Gene Symbol: GYG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015131770.2 | XP_014987256.1 | 788 | 109 | ||
| XM_015131771.2 | XP_014987257.1 | 801 | 109 | |||
| XM_015131772.2 | XP_014987258.1 | 788 | 109 | |||
| GenBank mRNA | JU334212.1 | - | 644 | 109 | ||
| JU334213.1 | - | 644 | 109 | |||
| JU471361.1 | - | 647 | 109 | |||
| JV046881.1 | - | 693 | 109 | |||
| JV601748.1 | - | 717 | 109 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

GYG1
Genomic Map