Assay Details
| Gene Symbol: C15H9orf64 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260888.1 | NP_001247817.1 | 699 | 90 | ||
| GenBank mRNA | JU322704.1 | - | 832 | 90 | ||
| JU473192.1 | - | 699 | 90 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C15H9orf64| Gene Symbol: C15H9orf64 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260888.1 | NP_001247817.1 | 699 | 90 | ||
| GenBank mRNA | JU322704.1 | - | 832 | 90 | ||
| JU473192.1 | - | 699 | 90 | |||