Assay Details
| Gene Symbol: FAM71F1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001090914.4 | XP_001090914.1 | 734 | 147 | ||
| XM_015134864.2 | XP_014990350.1 | 736 | 147 | |||
More Information
Set Membership: |
|

FAM71F1
Genomic Map