Assay Details
| Gene Symbol: ATOH8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015112777.2 | XP_014968263.2 | 1249 | 74 | ||
| XM_028832329.1 | XP_028688162.1 | 1251 | 74 | |||
| XM_028832330.1 | XP_028688163.1 | 1251 | 74 | |||
| GenBank mRNA | JV725521.1 | - | 129 | 74 | ||
More Information
Set Membership: |
|

ATOH8
Genomic Map