Assay Details
| Gene Symbol: PDGFA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028845418.1 | XP_028701251.1 | 1043 | 144 | ||
| XM_028845419.1 | XP_028701252.1 | 1043 | 144 | |||
| XM_028845420.1 | XP_028701253.1 | 529 | 144 | |||
| XM_028845421.1 | XP_028701254.1 | 726 | 144 | |||
| XM_028845422.1 | XP_028701255.1 | 529 | 144 | |||
| XM_028845423.1 | XP_028701256.1 | 693 | 144 | |||
More Information
Set Membership: |
|

PDGFA
Genomic Map