Assay Details
| Gene Symbol: FATE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261152.1 | NP_001248081.1 | 433 | 132 | ||
| XM_015128419.2 | XP_014983905.1 | 466 | 132 | |||
| GenBank mRNA | JV048619.1 | - | 433 | 132 | ||
| JV636441.1 | - | 432 | 132 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

FATE1
Genomic Map