Assay Details
| Gene Symbol: CDC42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015130151.2 | XP_014985637.1 | 649 | 102 | ||
| GenBank mRNA | JU337400.1 | - | 517 | 102 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CDC42| Gene Symbol: CDC42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015130151.2 | XP_014985637.1 | 649 | 102 | ||
| GenBank mRNA | JU337400.1 | - | 517 | 102 | ||
Set Membership: |
|