Assay Details
| Gene Symbol: ZBED9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001101327.4 | XP_001101327.2 | 1303 | 64 | ||
| XM_028847748.1 | XP_028703581.1 | 457 | 64 | |||
| GenBank mRNA | JV706286.1 | - | 694 | 64 | ||
More Information
Set Membership: |
|

ZBED9
Genomic Map