Assay Details
| Gene Symbol: EGLN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261091.1 | NP_001248020.1 | 1181 | 98 | ||
| GenBank mRNA | JU332747.1 | - | 1404 | 98 | ||
| JU474166.1 | - | 1398 | 98 | |||
| JV046075.1 | - | 1181 | 98 | |||
| JV671754.1 | - | 1420 | 98 | |||
| Gene Symbol: EGLN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028840215.1 | XP_028696048.1 | 1420 | 98 | ||
| GenBank mRNA | JV493305.1 | - | 302 | 98 | ||

EGLN2
Genomic Map