Assay Details
| Gene Symbol: CYP3A43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028844981.1 | XP_028700814.1 | 270 | 142 | ||
| XM_028844983.1 | XP_028700816.1 | 2536 | 142 | |||
| XM_028844985.1 | XP_028700818.1 | 208 | 142 | |||
| GenBank mRNA | EF589791.1 | - | 167 | 142 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_028844985.1 transcripts.
Set Membership: |
|

CYP3A43
Genomic Map