Assay Details
| Gene Symbol: CYP3A43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127630.1 | NP_001121102.1 | 529 | 144 | ||
| XM_028844981.1 | XP_028700814.1 | 779 | 144 | |||
| XM_028844982.1 | XP_028700815.1 | 2949 | 144 | |||
| XM_028844984.1 | XP_028700817.1 | 411 | 144 | |||
| XM_028844985.1 | XP_028700818.1 | 717 | 144 | |||
| GenBank mRNA | EF589792.1 | - | 529 | 144 | ||
More Information
Set Membership: |
|

CYP3A43
Genomic Map