Assay Details
| Gene Symbol: VNN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015137426.2 | XP_014992912.2 | 475 | 144 | ||
| XM_015137427.2 | XP_014992913.2 | 399 | 144 | |||
| XM_015137428.2 | XP_014992914.2 | 499 | 144 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

VNN2
Genomic Map