Assay Details
Gene Symbol: CREB3L4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265999.1 | NP_001252928.1 | 429 | 111 | ||
XM_015152617.2 | XP_015008103.1 | 1038 | 111 | |||
XM_028845397.1 | XP_028701230.1 | 404 | 111 | |||
GenBank mRNA | JV635973.1 | - | 429 | 111 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_028845397.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
