Assay Details
| Gene Symbol: ZNF689 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001111143.4 | XP_001111143.1 | 668 | 71 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF689| Gene Symbol: ZNF689 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001111143.4 | XP_001111143.1 | 668 | 71 | ||