Assay Details
| Gene Symbol: DHRS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194609.2 | NP_001181538.1 | 542 | 131 | ||
| XM_015143356.2 | XP_014998842.1 | 832 | 131 | |||
| GenBank mRNA | JU323104.1 | - | 555 | 131 | ||
| JU471900.1 | - | 562 | 131 | |||
| JV045445.1 | - | 558 | 131 | |||
| JV456141.1 | - | 467 | 131 | |||
| JV529938.1 | - | 515 | 131 | |||
More Information
Set Membership: |
|

DHRS1
Genomic Map