Assay Details
| Gene Symbol: TLCD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194451.2 | NP_001181380.1 | 297 | 115 | ||
| GenBank mRNA | JU325857.1 | - | 91 | 115 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TLCD1| Gene Symbol: TLCD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194451.2 | NP_001181380.1 | 297 | 115 | ||
| GenBank mRNA | JU325857.1 | - | 91 | 115 | ||
Set Membership: |
|