Assay Details
| Gene Symbol: LYPLAL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266865.1 | NP_001253794.1 | 484 | 127 | ||
| XM_015114967.2 | XP_014970453.1 | 864 | 127 | |||
| XM_028843342.1 | XP_028699175.1 | 727 | 127 | |||
| GenBank mRNA | JU332907.1 | - | 484 | 127 | ||
More Information
Set Membership: |
|

LYPLAL1
Genomic Map