Assay Details
| Gene Symbol: ARX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001091313.4 | XP_001091313.1 | 2701 | 122 | ||
| XM_028841937.1 | XP_028697770.1 | 2604 | 122 | |||
| XM_028841938.1 | XP_028697771.1 | 2512 | 122 | |||
More Information
Set Membership: |
|

ARX
Genomic Map