Assay Details
| Gene Symbol: PPIL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258155.1 | NP_001245084.1 | 874 | 131 | ||
| GenBank mRNA | JU326441.1 | - | 786 | 131 | ||
| JU472193.1 | - | 874 | 131 | |||
| JV048516.1 | - | 891 | 131 | |||
More Information
Set Membership: |
|

PPIL4
Genomic Map