Assay Details
| Gene Symbol: LGI4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015123549.2 | XP_014979035.1 | 1170 | 78 | ||
| XM_015123550.2 | XP_014979036.1 | 1174 | 78 | |||
| XM_015123552.2 | XP_014979038.1 | 1493 | 78 | |||
| XM_015123553.2 | XP_014979039.1 | 1102 | 78 | |||
| GenBank mRNA | JU326574.1 | - | 206 | 78 | ||
| JV453169.1 | - | 833 | 78 | |||
| JV565667.1 | - | 831 | 78 | |||
| JV667244.1 | - | 822 | 78 | |||
More Information
Set Membership: |
|

LGI4
Genomic Map