Assay Details
| Gene Symbol: CMTM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001084230.4 | XP_001084230.2 | 814 | 106 | ||
| XM_015126337.2 | XP_014981823.1 | 252 | 106 | |||
| GenBank mRNA | JV672662.1 | - | 286 | 106 | ||
More Information
Set Membership: |
|

CMTM3
Genomic Map