Assay Details
| Gene Symbol: NDUFB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001082389.4 | XP_001082389.2 | 339 | 71 | ||
| XM_028846385.1 | XP_028702218.1 | 494 | 71 | |||
| XM_028846386.1 | XP_028702219.1 | 578 | 71 | |||
| GenBank mRNA | JV582733.1 | - | 312 | 71 | ||
| JV617602.1 | - | 316 | 71 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

NDUFB2
Genomic Map