Assay Details
| Gene Symbol: LOC100430397 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002799391.3 | XP_002799437.2 | 83 | 96 | ||
| Gene Symbol: MRPL33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193419.1 | NP_001180348.1 | 45 | 96 | ||
| GenBank mRNA | CN804367.1 | - | 45 | 96 | ||
| JV490915.1 | - | 90 | 96 | |||
| JV601282.1 | - | 75 | 96 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001193419.1, XM_002799391.3 transcripts.
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

LOC100430397
Genomic Map