Assay Details
| Gene Symbol: SEC22B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261329.1 | NP_001248258.1 | 473 | 87 | ||
| GenBank mRNA | JU337220.1 | - | 483 | 87 | ||
| JU337221.1 | - | 483 | 87 | |||
| JU477935.1 | - | 482 | 87 | |||
| JV048776.1 | - | 473 | 87 | |||
| JV666213.1 | - | 485 | 87 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

SEC22B
Genomic Map