Assay Details
| Gene Symbol: TBX19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015125083.2 | XP_014980569.2 | 4149 | 94 | ||
| XM_015125090.2 | XP_014980576.2 | 798 | 94 | |||
| GenBank mRNA | JU496041.1 | - | 715 | 94 | ||
More Information
Set Membership: |
|

TBX19
Genomic Map