Assay Details
| Gene Symbol: GFER | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001082639.4 | XP_001082639.1 | 829 | 63 | ||
| XM_028841033.1 | XP_028696866.1 | 829 | 63 | |||
| XM_028841034.1 | XP_028696867.1 | 829 | 63 | |||
| GenBank mRNA | JU323653.1 | - | 307 | 63 | ||
| JU473389.1 | - | 328 | 63 | |||
| JV045729.1 | - | 327 | 63 | |||
| JV578316.1 | - | 333 | 63 | |||
| JV600563.1 | - | 338 | 63 | |||
More Information
Set Membership: |
|

GFER
Genomic Map