Assay Details
| Gene Symbol: LOC114676074 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028843022.1 | XP_028698855.1 | 126 | 124 | ||
| Gene Symbol: LAMA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001118617.4 | XP_001118617.4 | 2510 | 124 | ||
| GenBank mRNA | JV648837.1 | - | 2459 | 124 | ||
More Information
Set Membership: |
|

LOC114676074
Genomic Map