Assay Details
| Gene Symbol: LOC114676074 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028843022.1 | XP_028698855.1 | 1725 | 116 | ||
| Gene Symbol: LAMA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001118617.4 | XP_001118617.4 | 4109 | 116 | ||
| GenBank mRNA | JV058499.1 | - | 488 | 116 | ||
| JV648837.1 | - | 4058 | 116 | |||
More Information
Set Membership: |
|

LOC114676074
Genomic Map