Assay Details
| Gene Symbol: DNAL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260611.1 | NP_001247540.1 | 259 | 105 | ||
| GenBank mRNA | JU320327.1 | - | 293 | 105 | ||
| JU472484.1 | - | 227 | 105 | |||
| JV044998.1 | - | 259 | 105 | |||
| JV639432.1 | - | 305 | 105 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

DNAL4
Genomic Map