Assay Details
| Gene Symbol: VPS72 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266143.1 | NP_001253072.1 | 744 | 143 | ||
| XM_028843760.1 | XP_028699593.1 | 640 | 143 | |||
| GenBank mRNA | JU336121.1 | - | 755 | 143 | ||
| JV048077.1 | - | 744 | 143 | |||
| JV694692.1 | - | 756 | 143 | |||
More Information
Set Membership: |
|

VPS72
Genomic Map