Assay Details
| Gene Symbol: ECI2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001094776.4 | XP_001094776.1 | 772 | 91 | ||
| XM_015135494.2 | XP_014990980.1 | 756 | 91 | |||
| XM_028847871.1 | XP_028703704.1 | 861 | 91 | |||
| GenBank mRNA | JV527757.1 | - | 692 | 91 | ||
| JV705242.1 | - | 622 | 91 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

ECI2
Genomic Map