Assay Details
| Gene Symbol: TFPI | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195022.1 | NP_001181951.1 | 648 | 132 | ||
| GenBank mRNA | JV048080.1 | - | 759 | 132 | ||
| S73337.1 | - | 648 | 132 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

TFPI
Genomic Map