Assay Details
| Gene Symbol: ZKSCAN8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015135781.2 | XP_014991267.1 | 1426 | 92 | ||
| XM_015135782.2 | XP_014991268.1 | 1530 | 92 | |||
| XM_015135783.2 | XP_014991269.1 | 1424 | 92 | |||
| XM_028847761.1 | XP_028703594.1 | 589 | 92 | |||
| GenBank mRNA | JU322654.1 | - | 832 | 92 | ||
| JU471646.1 | - | 833 | 92 | |||
| JU471647.1 | - | 833 | 92 | |||
| JV519631.1 | - | 798 | 92 | |||
More Information
Set Membership: |
|

ZKSCAN8
Genomic Map