Assay Details
| Gene Symbol: IFI30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015123340.2 | XP_014978826.2 | 463 | 94 | ||
| GenBank mRNA | JU323483.1 | - | 371 | 94 | ||
| JU472042.1 | - | 380 | 94 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

IFI30
Genomic Map