Assay Details
| Gene Symbol: SLC16A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028842413.1 | XP_028698246.1 | 1398 | 77 | ||
| GenBank mRNA | JU328366.1 | - | 286 | 77 | ||
| JV626450.1 | - | 781 | 77 | |||
More Information
Set Membership: |
|

SLC16A2
Genomic Map