Assay Details
| Gene Symbol: MIA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028840218.1 | XP_028696051.1 | 1343 | 64 | ||
| XM_028840219.1 | XP_028696052.1 | 1116 | 64 | |||
| XM_028840220.1 | XP_028696053.1 | 1018 | 64 | |||
| Gene Symbol: LOC702844 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015123966.2 | XP_014979452.1 | 1019 | 64 | ||
| XM_015123967.2 | XP_014979453.1 | 1117 | 64 | |||
| XM_028842807.1 | XP_028698640.1 | 782 | 64 | |||
| GenBank mRNA | JV635667.1 | - | 501 | 64 | ||
More Information
Set Membership: |
|

MIA
Genomic Map